3-Chloro-2-nitropyridine ≥97%

Supplier: Thermo Scientific
Warning

L19863.03 L19863.06 L19863.MD
ALFAL19863.03EA 283 SGD
ALFAL19863.03 ALFAL19863.06 ALFAL19863.MD
3-Chloro-2-nitropyridine ≥97%
3-Chloro-2-nitropyridine

Purity: 97% CAS No.: 54231-32-2 Molecular Formula: C5H3ClN2O2 Formula Weight: 158.54 Melting Point: 89-92° Beilstein Reference: 1637069

Formula: C₅H₃ClN₂O₂
Melting Pt: 89…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00490110
CAS Number: 54231-32-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR