4-Nitro-3-picoline-N-oxide ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
4-Nitropicoline N-oxide, 3-Methyl-4-nitropyridine 1-oxide, 3-Methyl-4-nitropyridine-N-oxide, 3-Methyl-4-nitropyridine N-oxide, 4-Nitro-3-picoline N-oxide, 3-Methyl-4-nitropyridine-1-oxide
Purity: 98%
CAS No.: 1074-98-2
Molecular Formula: C6H6N2O3
Formula Weight: 154.13
Melting Point: 134-138°
Beilstein Reference: 140157
UN No.: 2811
MDL No.: MFCD00014626
Formula:
C₆H₆N₂O₃ MW: 154,13 g/mol Melting Pt: 134…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014626 CAS Number: 1074-98-2 EINECS: 214-050-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...