Cleanroom gloves, nitrile, Kimtech™ G6
Supplier: KIMBERLY CLARK
68700 68701 68702 68703 68704
KIMB68700EA 144 SGD
KIMB68700 KIMB68701 KIMB68702 KIMB68703 KIMB68704
Cleanroom gloves, nitrile, Kimtech™ G6
Gloves
Ambidextrous cleanroom gloves feature textured fingertips for precision work in pharmaceutical, biotech, semiconductor, medical device, data storage, and touch panel applications.
  • Non-sterile, powder-free nitrile gloves
  • AQL 1,5
  • 5,1 mil/0,13mm thickness at fingertips
  • Double-bagged for cleanroom use
  • Accelerator-, DOP-, and amide-free
  • Silicone-free material
  • Beaded cuff for easy donning
Ideal for use in pharma grade C&D, ISO 6 - 8, and Class 1000 cleanrooms.
Packaging: 100 gloves per bag, 1000 gloves per case.