Polyethylene glycol 3,350, Biotechnology Grade

Supplier: VWR Chemicals

Synonyms: PEG 3,350, PEG 3350

VWRC0955-1KGEA 139 SGD
VWRC0955-1KG VWRC0955-500G
Polyethylene glycol 3,350, Biotechnology Grade
Polyethylene glycol 3,350
Formula: H(OCH₂CH₂)nOH
MW: 3350 g/mol
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR