2-Propanol ≥99.9%, Finyte®, J.T.Baker®
Supplier: Avantor
|
Danger
|
Synonyms:
Isopropyl rubbing alcohol, Isopropanol, 2-Hydroxy propane, IPA, Dimethyl carbinol, Isopropyl alcohol, Propan-2-ol
BAKR5978-89EA
1930
SGD
BAKR5978-89
BAKR5978-99
2-Propanol ≥99.9%, Finyte®, J.T.Baker®
2-Propanol
Formula:
(CH₃)₂CHOH MW: 60,1 g/mol Boiling Pt: 82 °C (1013 hPa) Melting Pt: –89 °C Density: 0,786 g/cm³ (20 °C) Flash Pt: 12 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011674 CAS Number: 67-63-0 EINECS: 200-661-7 UN: 1219 ADR: 3,II REACH: 01-2119457558-25 |
Specification Test Results
For Microelectronic Use | |
Assay (CH₃CHOHCH₃) | ≥99.9 % |
Color (APHA) | ≤6 |
Residue after Evaporation | ≤3000.0 ppb |
Solubility in H₂O | Passes Test |
Water (H₂O)(by Karl Fischer titrn) | ≤0.05 % |
Acidity (µeq/g) | ≤0.2 |
Alkalinity (µeq/g) | ≤0.1 |
Chloride (Cl) | ≤60.0 ppb |
Nitrate (NO₃) | ≤0.1 ppm |
Sulfate (SO₄) | ≤0.1 ppm |
Phosphate (PO₄) | ≤75.0 ppb |
Trace Impurities - Aluminum (Al) | ≤10.0 ppb |
Arsenic and Antimony (as As) | ≤10.0 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤10.0 ppb |
Trace Impurities - Boron (B) | ≤3.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤10.0 ppb |
Trace Impurities - Calcium (Ca) | ≤10.0 ppb |
Trace Impurities - Chromium (Cr) | ≤10.0 ppb |
Trace Impurities - Cobalt (Co) | ≤10.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤10.0 ppb |
Trace Impurities - Gold (Au) | ≤5.0 ppb |
Trace Impurities - Iron (Fe) | ≤5.0 ppb |
Trace Impurities - Lead (Pb) | ≤10.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤10.0 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Molybdenum (Mo) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤10.0 ppb |
Trace Impurities - Niobium (Nb) | ≤10.0 ppb |
Trace Impurities - Potassium (K) | ≤10.0 ppb |
Trace Impurities - Silicon (Si) | ≤10.0 ppb |
Trace Impurities - Silver (Ag) | ≤5.0 ppb |
Trace Impurities - Sodium (Na) | ≤10.0 ppb |
Trace Impurities - Strontium (Sr) | ≤10.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤10.0 ppb |
Trace Impurities - Tin (Sn) | ≤10.0 ppb |
Trace Impurities - Titanium (Ti) | ≤10.0 ppb |
Trace Impurities - Vanadium (V) | ≤10.0 ppb |
Trace Impurities - Zinc (Zn) | ≤10.0 ppb |
Trace Impurities - Zirconium (Zr) | ≤10.0 ppb |
Particle Count - 0.5 µm and greater | ≤150 par/ml |
Particle Count - 1.0 µm and greater | ≤25 par/ml |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...