Acetone ≥99.5%, CMOS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

BAKR5965-05EA 27.9 SGD
BAKR5965-05 BAKR5965-16
Acetone ≥99.5%, CMOS, J.T.Baker®
Acetone
Formula: CH₃COCH₃
MW: 58,08 g/mol
Flash Pt: <–20 °C
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
UN: 1090
ADR: 3,II

Order Now

Specification Test Results

For Microelectronic Use
Assay ((CH₃)₂CO) (by GC, corrected for water) ≥99.5 %
Color (APHA) ≤10
Specific Gravity at 20°/20°C 0.790 - 0.793
Residue after Evaporation ≤5 ppm
Titrable Acid (µeq/g) ≤0.3
Titrable Base (µeq/g) ≤0.5
Water (H₂O) ≤0.5 %
Solubility in H₂O Passes Test
Chloride (Cl) ≤0.2 ppm
Phosphate (PO₄) ≤0.05 ppm
Trace Impurities - Aluminum (Al) ≤50.0 ppb
Arsenic and Antimony (as As) ≤5.0 ppb
Trace Impurities - Barium (Ba) ≤20.0 ppb
Trace Impurities - Beryllium (Be) ≤10.0 ppb
Trace Impurities - Bismuth (Bi) ≤20.0 ppb
Trace Impurities - Boron (B) ≤10.0 ppb
Trace Impurities - Cadmium (Cd) ≤10.0 ppb
Trace Impurities - Calcium (Ca) ≤25.0 ppb
Trace Impurities - Chromium (Cr) ≤10.0 ppb
Trace Impurities - Cobalt (Co) ≤10.0 ppb
Trace Impurities - Copper (Cu) ≤10.0 ppb
Trace Impurities - Gallium (Ga) ≤10.0 ppb
Trace Impurities - Germanium (Ge) ≤10.0 ppb
Trace Impurities - Gold (Au) ≤20 ppb
Trace Impurities - Iron (Fe) ≤20.0 ppb
Trace Impurities - Lead (Pb) ≤10.0 ppb
Trace Impurities - Lithium (Li) ≤10.0 ppb
Trace Impurities - Magnesium (Mg) ≤20 ppb
Trace Impurities - Manganese (Mn) ≤10.0 ppb
Trace Impurities - Molybdenum (Mo) ≤10.0 ppb
Trace Impurities - Nickel (Ni) ≤10.0 ppb
Trace Impurities - Niobium (Nb) ≤50.0 ppb
Trace Impurities - Potassium (K) ≤10.0 ppb
Trace Impurities - Silicon (Si) ≤50 ppb
Trace Impurities - Silver (Ag) ≤10.0 ppb
Trace Impurities - Sodium (Na) ≤10.0 ppb
Trace Impurities - Strontium (Sr) ≤10.0 ppb
Trace Impurities - Tantalum (Ta) ≤50.0 ppb
Trace Impurities - Thallium (Tl) ≤10.0 ppb
Trace Impurities - Tin (Sn) ≤20.0 ppb
Trace Impurities - Titanium (Ti) ≤10.0 ppb
Trace Impurities - Vanadium (V) ≤10.0 ppb
Trace Impurities - Zinc (Zn) ≤20.0 ppb
Trace Impurities - Zirconium (Zr) ≤10.0 ppb
Particle Count - 0.5 µm and greater ≤100 par/ml
Particle Count - 1.0 µm and greater ≤8 par/ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR