Orthophosphoric acid ≥85%, BAKER ANALYZED® ACS, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: Phosphoric acid

Management of Change (MOC) category = N

BAKR0255-80EA 70.7 SGD
BAKR0255-80 BAKR0255-88
Orthophosphoric acid ≥85%, BAKER ANALYZED® ACS, J.T.Baker®
Orthophosphoric acid
Formula: H₃PO₄
MW: 98 g/mol
MDL Number: MFCD00011340
CAS Number: 7664-38-2
UN: 1805
ADR: 8,III

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (H₃PO₄) (by acidimetry) 85.0 - 87.0 %
Calcium (Ca) ≤0.002 %
Color (APHA) ≤10
Insoluble Matter ≤0.001 %
ACS - Magnesium (Mg) ≤0.002 %
Sulfate (SO₄) ≤12 ppm
Volatile Acids (as CH₃COOH) ≤0.001 %
Reducing Substances Passes Test
Chloride (Cl) ≤3 ppm
Nitrate (NO₃) ≤5 ppm
Trace Impurities - Antimony (Sb) ≤20.000 ppm
Trace Impurities - Arsenic (As) ≤0.500 ppm
Trace Impurities - Iron (Fe) ≤10 ppm
Heavy Metals (as Pb) ≤8 ppm
Trace Impurities - Manganese (Mn) ≤0.500 ppm
Trace Impurities - Potassium (K) ≤40.000 ppm
Trace Impurities - Sodium (Na) ≤50.000 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR