2-Methylbutane ≥99.0%, J.T.Baker®

Supplier: Avantor
Danger

Synonyms: iso-Pentane

Management of Change (MOC) category = N

BAKRQ223-07EA 108 SGD
BAKRQ223-07 BAKRQ223-08
2-Methylbutane ≥99.0%, J.T.Baker®
2-Methylbutane
Formula: CH₃CH₂CH(CH₃)₂
MW: 72,15 g/mol
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0,62 g/cm³ (20 °C)
Flash Pt: –51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Store at or below 68°F (20°C).
Assay ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR