HPLC columns, HALO® Phenyl-Hexyl
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-106 91812-206 91812-306 91812-406 91812-506 91812-606 91812-706 91812-906 91813-106 91813-206 91813-306 91813-406 91813-506 91813-606 91813-706 91813-906 92810-406 92810-606 92810-706 92811-406 92811-606 92811-706 92812-106 92812-206 92812-306 92812-406 92812-506 92812-606 92812-706 92812-906 92813-106 92813-206 92813-306 92813-406 92813-506 92813-606 92813-706 92813-906 92814-106 92814-206 92814-306 92814-406 92814-506 92814-606 92814-706 92814-906 95810-406 95810-606 95810-706 95810-906 95812-106 95812-206 95812-306 95812-406 95812-506 95812-606 95812-706 95812-906 95813-106 95813-206 95813-306 95813-406 95813-506 95813-606 95813-706 95813-906 95814-106 95814-206 95814-306 95814-406 95814-506 95814-606 95814-706 95814-906 98215-406 98215-606 98215-706 98216-406 98216-606 98216-706
AMTE91812-106EA 1510 SGD
AMTE91812-106 AMTE91812-206 AMTE91812-306 AMTE91812-406 AMTE91812-506 AMTE91812-606 AMTE91812-706 AMTE91812-906 AMTE91813-106 AMTE91813-206 AMTE91813-306 AMTE91813-406 AMTE91813-506 AMTE91813-606 AMTE91813-706 AMTE91813-906 AMTE92810-406 AMTE92810-606 AMTE92810-706 AMTE92811-406 AMTE92811-606 AMTE92811-706 AMTE92812-106 AMTE92812-206 AMTE92812-306 AMTE92812-406 AMTE92812-506 AMTE92812-606 AMTE92812-706 AMTE92812-906 AMTE92813-106 AMTE92813-206 AMTE92813-306 AMTE92813-406 AMTE92813-506 AMTE92813-606 AMTE92813-706 AMTE92813-906 AMTE92814-106 AMTE92814-206 AMTE92814-306 AMTE92814-406 AMTE92814-506 AMTE92814-606 AMTE92814-706 AMTE92814-906 AMTE95810-406 AMTE95810-606 AMTE95810-706 AMTE95810-906 AMTE95812-106 AMTE95812-206 AMTE95812-306 AMTE95812-406 AMTE95812-506 AMTE95812-606 AMTE95812-706 AMTE95812-906 AMTE95813-106 AMTE95813-206 AMTE95813-306 AMTE95813-406 AMTE95813-506 AMTE95813-606 AMTE95813-706 AMTE95813-906 AMTE95814-106 AMTE95814-206 AMTE95814-306 AMTE95814-406 AMTE95814-506 AMTE95814-606 AMTE95814-706 AMTE95814-906 AMTE98215-406 AMTE98215-606 AMTE98215-706 AMTE98216-406 AMTE98216-606 AMTE98216-706
HPLC columns, HALO® Phenyl-Hexyl
Chromatography Columns
HALO® Phenyl-Hexyl columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å Phenyl-Hexyl columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for separating aromatic compounds using pi-pi interactions
  • Alternate selectivity to alkyl phases
  • Also available with 160 Å pores for peptide mapping
  • Particle Sizes: 2, 2,7 or 5 μm
More Product Information