HPLC columns, HALO® PFP
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-109 91812-209 91812-309 91812-409 91812-509 91812-609 91812-709 91812-909 91813-109 91813-209 91813-309 91813-409 91813-509 91813-609 91813-709 91813-909 92810-409 92810-609 92810-709 92811-409 92811-609 92811-709 92812-109 92812-209 92812-309 92812-409 92812-509 92812-609 92812-709 92812-909 92813-109 92813-209 92813-309 92813-409 92813-509 92813-609 92813-709 92813-909 92814-109 92814-209 92814-309 92814-409 92814-509 92814-609 92814-709 92814-909 95810-409 95810-609 95810-709 95810-909 95812-109 95812-209 95812-309 95812-409 95812-509 95812-609 95812-709 95812-909 95813-109 95813-209 95813-309 95813-409 95813-509 95813-609 95813-709 95813-909 95814-109 95814-209 95814-309 95814-409 95814-509 95814-609 95814-709 95814-909 98215-409 98215-609 98215-709 98216-409 98216-609 98216-709
AMTE91812-109EA 861 SGD
AMTE91812-109 AMTE91812-209 AMTE91812-309 AMTE91812-409 AMTE91812-509 AMTE91812-609 AMTE91812-709 AMTE91812-909 AMTE91813-109 AMTE91813-209 AMTE91813-309 AMTE91813-409 AMTE91813-509 AMTE91813-609 AMTE91813-709 AMTE91813-909 AMTE92810-409 AMTE92810-609 AMTE92810-709 AMTE92811-409 AMTE92811-609 AMTE92811-709 AMTE92812-109 AMTE92812-209 AMTE92812-309 AMTE92812-409 AMTE92812-509 AMTE92812-609 AMTE92812-709 AMTE92812-909 AMTE92813-109 AMTE92813-209 AMTE92813-309 AMTE92813-409 AMTE92813-509 AMTE92813-609 AMTE92813-709 AMTE92813-909 AMTE92814-109 AMTE92814-209 AMTE92814-309 AMTE92814-409 AMTE92814-509 AMTE92814-609 AMTE92814-709 AMTE92814-909 AMTE95810-409 AMTE95810-609 AMTE95810-709 AMTE95810-909 AMTE95812-109 AMTE95812-209 AMTE95812-309 AMTE95812-409 AMTE95812-509 AMTE95812-609 AMTE95812-709 AMTE95812-909 AMTE95813-109 AMTE95813-209 AMTE95813-309 AMTE95813-409 AMTE95813-509 AMTE95813-609 AMTE95813-709 AMTE95813-909 AMTE95814-109 AMTE95814-209 AMTE95814-309 AMTE95814-409 AMTE95814-509 AMTE95814-609 AMTE95814-709 AMTE95814-909 AMTE98215-409 AMTE98215-609 AMTE98215-709 AMTE98216-409 AMTE98216-609 AMTE98216-709
HPLC columns, HALO® PFP
Chromatography Columns
HALO® PFP columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å PFP columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for aromatics and electron-rich compounds
  • Alternate selectivity to alkyl phases
  • Useful in reversed-phase and HILIC modes
  • Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.