HPLC columns HALO® RP-Amide
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-107 91812-207 91812-307 91812-407 91812-507 91812-607 91812-707 91812-907 91813-107 91813-207 91813-307 91813-407 91813-507 91813-607 91813-707 91813-907 92810-407 92810-607 92810-707 92811-407 92811-607 92811-707 92812-107 92812-207 92812-307 92812-407 92812-507 92812-607 92812-707 92812-907 92813-107 92813-207 92813-307 92813-407 92813-507 92813-607 92813-707 92813-907 92814-107 92814-207 92814-307 92814-407 92814-507 92814-607 92814-707 92814-907 95810-407 95810-607 95810-707 95810-907 95812-107 95812-207 95812-307 95812-407 95812-507 95812-607 95812-707 95812-907 95813-107 95813-207 95813-307 95813-407 95813-507 95813-607 95813-707 95813-907 95814-107 95814-207 95814-307 95814-407 95814-507 95814-607 95814-707 95814-907 98215-407 98215-607 98215-707 98216-407 98216-607 98216-707
AMTE91812-107EA 1510 SGD
AMTE91812-107 AMTE91812-207 AMTE91812-307 AMTE91812-407 AMTE91812-507 AMTE91812-607 AMTE91812-707 AMTE91812-907 AMTE91813-107 AMTE91813-207 AMTE91813-307 AMTE91813-407 AMTE91813-507 AMTE91813-607 AMTE91813-707 AMTE91813-907 AMTE92810-407 AMTE92810-607 AMTE92810-707 AMTE92811-407 AMTE92811-607 AMTE92811-707 AMTE92812-107 AMTE92812-207 AMTE92812-307 AMTE92812-407 AMTE92812-507 AMTE92812-607 AMTE92812-707 AMTE92812-907 AMTE92813-107 AMTE92813-207 AMTE92813-307 AMTE92813-407 AMTE92813-507 AMTE92813-607 AMTE92813-707 AMTE92813-907 AMTE92814-107 AMTE92814-207 AMTE92814-307 AMTE92814-407 AMTE92814-507 AMTE92814-607 AMTE92814-707 AMTE92814-907 AMTE95810-407 AMTE95810-607 AMTE95810-707 AMTE95810-907 AMTE95812-107 AMTE95812-207 AMTE95812-307 AMTE95812-407 AMTE95812-507 AMTE95812-607 AMTE95812-707 AMTE95812-907 AMTE95813-107 AMTE95813-207 AMTE95813-307 AMTE95813-407 AMTE95813-507 AMTE95813-607 AMTE95813-707 AMTE95813-907 AMTE95814-107 AMTE95814-207 AMTE95814-307 AMTE95814-407 AMTE95814-507 AMTE95814-607 AMTE95814-707 AMTE95814-907 AMTE98215-407 AMTE98215-607 AMTE98215-707 AMTE98216-407 AMTE98216-607 AMTE98216-707
HPLC columns HALO® RP-Amide
Chromatography Columns
HALO® RP-Amide columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO 90 Å RP-Amide columns are optimized for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for basic compounds (alcohols, acids, phenols, catechins)
  • Alternate selectivity to alkyl phases
  • 100% aqueous compatible
  • Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable
Endcapped Yes
pH range 2 - 9
Temperature limit 60 ⁰C (pH 2); 40 ⁰C (pH 9)
USP code L60