HPLC columns, HALO® AQ-C18
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-122 91812-222 91812-322 91812-422 91812-522 91812-622 91812-722 91812-922 91813-122 91813-222 91813-322 91813-422 91813-522 91813-622 91813-722 91813-922 92810-422 92810-622 92810-722 92811-422 92811-622 92811-722 92812-122 92812-222 92812-322 92812-422 92812-522 92812-622 92812-722 92812-922 92813-122 92813-222 92813-322 92813-422 92813-522 92813-622 92813-722 92813-922 92814-122 92814-222 92814-322 92814-422 92814-522 92814-622 92814-722 92814-922 95810-422 95810-622 95810-722 95810-922 95812-122 95812-222 95812-322 95812-422 95812-522 95812-622 95812-722 95812-922 95813-122 95813-222 95813-322 95813-422 95813-522 95813-622 95813-722 95813-922 95814-122 95814-222 95814-322 95814-422 95814-522 95814-622 95814-722 95814-922 98215-422 98215-622 98215-722 98216-422 98216-622 98216-722
AMTE91812-122EA 1510 SGD
AMTE91812-122 AMTE91812-222 AMTE91812-322 AMTE91812-422 AMTE91812-522 AMTE91812-622 AMTE91812-722 AMTE91812-922 AMTE91813-122 AMTE91813-222 AMTE91813-322 AMTE91813-422 AMTE91813-522 AMTE91813-622 AMTE91813-722 AMTE91813-922 AMTE92810-422 AMTE92810-622 AMTE92810-722 AMTE92811-422 AMTE92811-622 AMTE92811-722 AMTE92812-122 AMTE92812-222 AMTE92812-322 AMTE92812-422 AMTE92812-522 AMTE92812-622 AMTE92812-722 AMTE92812-922 AMTE92813-122 AMTE92813-222 AMTE92813-322 AMTE92813-422 AMTE92813-522 AMTE92813-622 AMTE92813-722 AMTE92813-922 AMTE92814-122 AMTE92814-222 AMTE92814-322 AMTE92814-422 AMTE92814-522 AMTE92814-622 AMTE92814-722 AMTE92814-922 AMTE95810-422 AMTE95810-622 AMTE95810-722 AMTE95810-922 AMTE95812-122 AMTE95812-222 AMTE95812-322 AMTE95812-422 AMTE95812-522 AMTE95812-622 AMTE95812-722 AMTE95812-922 AMTE95813-122 AMTE95813-222 AMTE95813-322 AMTE95813-422 AMTE95813-522 AMTE95813-622 AMTE95813-722 AMTE95813-922 AMTE95814-122 AMTE95814-222 AMTE95814-322 AMTE95814-422 AMTE95814-522 AMTE95814-622 AMTE95814-722 AMTE95814-922 AMTE98215-422 AMTE98215-622 AMTE98215-722 AMTE98216-422 AMTE98216-622 AMTE98216-722
HPLC columns, HALO® AQ-C18
Chromatography Columns
HALO® AQ-C18 columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å AQ-C18 columns are optimised for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Ideal for mixtures of polar and non-polar solutes
  • High retentivity of C18 with alternate selectivity
  • 100% aqueous compatible
  • Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core',
More Product Information