HPLC columns, HALO® C18
Supplier: ADVANCED MATERIALS TECHNOLOGY
91812-102 91812-202 91812-302 91812-402 91812-502 91812-602 91812-702 91812-902 91813-102 91813-202 91813-302 91813-402 91813-502 91813-602 91813-702 91813-902 92810-402 92810-602 92810-702 92811-402 92811-602 92811-702 92812-102 92812-202 92812-302 92812-402 92812-502 92812-602 92812-702 92812-902 92813-102 92813-202 92813-302 92813-402 92813-502 92813-602 92813-702 92813-902 92814-102 92814-202 92814-302 92814-402 92814-502 92814-602 92814-702 92814-902 95810-402 95810-602 95810-702 95810-902 95812-102 95812-202 95812-302 95812-402 95812-502 95812-602 95812-702 95812-902 95813-102 95813-202 95813-302 95813-402 95813-502 95813-602 95813-702 95813-902 95814-102 95814-202 95814-302 95814-402 95814-502 95814-602 95814-702 95814-902 98215-402 98215-602 98215-702 98216-402 98216-602 98216-702
AMTE91812-102EA 861 SGD
AMTE91812-102 AMTE91812-202 AMTE91812-302 AMTE91812-402 AMTE91812-502 AMTE91812-602 AMTE91812-702 AMTE91812-902 AMTE91813-102 AMTE91813-202 AMTE91813-302 AMTE91813-402 AMTE91813-502 AMTE91813-602 AMTE91813-702 AMTE91813-902 AMTE92810-402 AMTE92810-602 AMTE92810-702 AMTE92811-402 AMTE92811-602 AMTE92811-702 AMTE92812-102 AMTE92812-202 AMTE92812-302 AMTE92812-402 AMTE92812-502 AMTE92812-602 AMTE92812-702 AMTE92812-902 AMTE92813-102 AMTE92813-202 AMTE92813-302 AMTE92813-402 AMTE92813-502 AMTE92813-602 AMTE92813-702 AMTE92813-902 AMTE92814-102 AMTE92814-202 AMTE92814-302 AMTE92814-402 AMTE92814-502 AMTE92814-602 AMTE92814-702 AMTE92814-902 AMTE95810-402 AMTE95810-602 AMTE95810-702 AMTE95810-902 AMTE95812-102 AMTE95812-202 AMTE95812-302 AMTE95812-402 AMTE95812-502 AMTE95812-602 AMTE95812-702 AMTE95812-902 AMTE95813-102 AMTE95813-202 AMTE95813-302 AMTE95813-402 AMTE95813-502 AMTE95813-602 AMTE95813-702 AMTE95813-902 AMTE95814-102 AMTE95814-202 AMTE95814-302 AMTE95814-402 AMTE95814-502 AMTE95814-602 AMTE95814-702 AMTE95814-902 AMTE98215-402 AMTE98215-602 AMTE98215-702 AMTE98216-402 AMTE98216-602 AMTE98216-702
HPLC columns, HALO® C18
Chromatography Columns
HALO® C18 columns made with innovative Fused-Core® particle technology for excellent reproducibility and column lifetimes provide fast, high resolution separations. Available in three particle sizes and a wide range of column dimensions, HALO® 90 Å C18 columns are optimised for high performance in HPLC, UHPLC, and LCMS small molecule applications.
  • Universal phase for acids, bases and neutral solutes with a wide range of hydrophobicities
  • Excellent stability at low to mid mobile phase pH
  • Widely published and most utilised stationary phase
  • Particle sizes: 2, 2,7 or 5 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® guard columns are available for this phase.