HPLC columns, HALO® ES-C18, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
91122-102 91122-202 91122-302 91122-402 91122-502 91122-602 91122-702 91122-902 91123-102 91123-302 91123-402 91123-502 91123-602 91123-702 91123-902 91225-402 91225-602 91225-702 91226-402 91226-602 91226-702 92120-402 92120-602 92120-702 92121-402 92121-602 92121-702 92121-902 92122-102 92122-202 92122-302 92122-402 92122-502 92122-602 92122-702 92122-902 92123-102 92123-202 92123-302 92123-402 92123-502 92123-602 92123-702 92123-902 92124-102 92124-202 92124-302 92124-402 92124-502 92124-602 92124-702 92124-902 95120-402 95120-602 95120-702 95120-902 95122-102 95122-202 95122-302 95122-402 95122-502 95122-602 95122-702 95122-902 95123-102 95123-202 95123-302 95123-402 95123-502 95123-602 95123-702 95123-902 95124-102
AMTE91122-102EA 1510 SGD
AMTE91122-102 AMTE91122-202 AMTE91122-302 AMTE91122-402 AMTE91122-502 AMTE91122-602 AMTE91122-702 AMTE91122-902 AMTE91123-102 AMTE91123-302 AMTE91123-402 AMTE91123-502 AMTE91123-602 AMTE91123-702 AMTE91123-902 AMTE91225-402 AMTE91225-602 AMTE91225-702 AMTE91226-402 AMTE91226-602 AMTE91226-702 AMTE92120-402 AMTE92120-602 AMTE92120-702 AMTE92121-402 AMTE92121-602 AMTE92121-702 AMTE92121-902 AMTE92122-102 AMTE92122-202 AMTE92122-302 AMTE92122-402 AMTE92122-502 AMTE92122-602 AMTE92122-702 AMTE92122-902 AMTE92123-102 AMTE92123-202 AMTE92123-302 AMTE92123-402 AMTE92123-502 AMTE92123-602 AMTE92123-702 AMTE92123-902 AMTE92124-102 AMTE92124-202 AMTE92124-302 AMTE92124-402 AMTE92124-502 AMTE92124-602 AMTE92124-702 AMTE92124-902 AMTE95120-402 AMTE95120-602 AMTE95120-702 AMTE95120-902 AMTE95122-102 AMTE95122-202 AMTE95122-302 AMTE95122-402 AMTE95122-502 AMTE95122-602 AMTE95122-702 AMTE95122-902 AMTE95123-102 AMTE95123-202 AMTE95123-302 AMTE95123-402 AMTE95123-502 AMTE95123-602 AMTE95123-702 AMTE95123-902 AMTE95124-102
HPLC columns, HALO® ES-C18, BIOCLASS
Chromatography Columns
HALO® BIOCLASS Phenyl-Hexyl peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast and ultrahigh resolution separations of peptides and polypeptides up to 20 kDa.
  • Ideal for tryptic digests, post-translational modifications, (PTM)s; polypeptides
  • HALO® Peptide columns can be used in series to increase peak capacity for UHPLC and HPLC analyses of complex tryptic digest samples
  • Particle Size: 2,7 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® peptide columns offer comparable peakMore Product Information