HPLC columns, HALO® Phenyl-Hexyl, BIOCLASS
Supplier: ADVANCED MATERIALS TECHNOLOGY
91215-406 91215-606 91215-706 91216-406 91216-606 91216-706 92110-406 92110-606 92110-706 92111-406 92111-606 92111-706 92112-106 92112-206 92112-306 92112-406 92112-506 92112-606 92112-706 92112-906 92113-106 92113-206 92113-306 92113-406 92113-506 92113-606 92113-706 92113-906 92114-106 92114-206 92114-306 92114-406 92114-506 92114-606 92114-706 92114-906
AMTE91215-406EA 2400 SGD
AMTE91215-406 AMTE91215-606 AMTE91215-706 AMTE91216-406 AMTE91216-606 AMTE91216-706 AMTE92110-406 AMTE92110-606 AMTE92110-706 AMTE92111-406 AMTE92111-606 AMTE92111-706 AMTE92112-106 AMTE92112-206 AMTE92112-306 AMTE92112-406 AMTE92112-506 AMTE92112-606 AMTE92112-706 AMTE92112-906 AMTE92113-106 AMTE92113-206 AMTE92113-306 AMTE92113-406 AMTE92113-506 AMTE92113-606 AMTE92113-706 AMTE92113-906 AMTE92114-106 AMTE92114-206 AMTE92114-306 AMTE92114-406 AMTE92114-506 AMTE92114-606 AMTE92114-706 AMTE92114-906
HPLC columns, HALO® Phenyl-Hexyl, BIOCLASS
Chromatography Columns
HALO® BIOCLASS Phenyl-Hexyl peptide columns made with innovative Fused-Core® particle technology are ideal for both ultrafast and ultrahigh resolution separations of peptides and polypeptides up to 20 kDa.
  • Ideal for tryptic digests, post-translational modifications, (PTM)s; polypeptides
  • HALO® Peptide columns can be used in series to increase peak capacity for UHPLC and HPLC analyses of complex tryptic digest samples
  • Particle size: 2,7 μm superficially porous particle silica ('solid-core', 'core-shell')
HALO® peptide columns offer comparable peakMore Product Information