Cleanroom gloves, nitrile, Kimtech™ G3 EvT
Supplier: KIMBERLY CLARK
38701 38702 38700 38703 38704
KIMB38701EA 285 SGD
KIMB38701 KIMB38702 KIMB38700 KIMB38703 KIMB38704
Cleanroom gloves, nitrile, Kimtech™ G3 EvT
Gloves
Kimtech™ G3 EvT nitrile gloves are designed with ‘Extra-Value Technology’ to have low particles and extractables. They are cleaned to meet the needs of a critical cleanroom environment.
  • Textured and tacky surface for increased dexterity, with beaded cuffs for easy donning
  • Low particles and extractables
  • Durable and powder-free
  • Not made with silicone or natural rubber latex
Gloves have excellent overall tactile performance. Additionally, the textured and tacky surface provides increased dexterity for critical tasks in semiconductor, electronics, andMore Product Information