Cut resistant gloves, JACKSON SAFETY* G60, Level 3
Supplier: KIMBERLY CLARK
13823 13824 13825
KIMB13823EA 0 SGD
KIMB13823 KIMB13824 KIMB13825
Cut resistant gloves, JACKSON SAFETY* G60, Level 3
Gloves
A range of cost effective, durable gloves and sleeves suitable for processes where there is cutting or laceration risk to workers hands and arms.
  • Latex free
  • Available in different sizes with colour coded cuff
  • Polyurethane coating offering good dexterity, dry and oil grip
These gloves are suitable for metal fabrication, glass and sharp objects handling, machinery and bending
small parts.
Level 4 abrasion resistance with high breathability as material transmits heat away. Durable and comfortable.
Certifications: Level 3 cut resistance (EN 388:4342).More Product Information