Examination gloves, Kimtech™ Purple Nitrile-Xtra™
Supplier: KIMBERLY CLARK
50601M 50603M 50604M
KIMB50601MEA 0 SGD
KIMB50601M KIMB50603M KIMB50604M
Examination gloves, Kimtech™ Purple Nitrile-Xtra™
Gloves
These purple nitrile gloves guard against incidental splashes, are made of a highly durable nitrile polymer and are powder free and not made with natural rubber latex (some say 'latex free').
  • Non sterile, powder-free, textured fingertips
  • Excellent dexterity and tactile sensitivity
  • Comfortable fit
  • Recyclable through the RightCycle program, so the gloves won't end up in a landfill
These comfortable gloves are cleared for use in chemotherapy.
Certifications: AQL 1.0 (exceeds the ASTM 2.5 standard for critical defects or pinholes).
Packaging: 50 gloves/box.