VWR®, PCR Tubes, Classic
Supplier: VWR Collection
VWRA732-3649EA 145 SGD
VWRA732-3649 VWRA732-3650 VWRA732-3651 VWRA732-3652 VWRA732-3653 VWRA732-3654 VWRA732-3655 VWRA732-3656
VWR®, PCR Tubes, Classic
Tubes PCR Tubes
VWR® PCR Classic plasticwares are DNAse-, RNAse- and endotoxin-free for hassle free application outcome in PCR labs.
  • Ultrathin wall helps with superior heat transmission
  • Sterilised as per SAL 10⁻⁶ gamma irradiation standard
Tubes and strips are made of virgin PP and the manufacturing facility is compliant to 100K cleanroom as well as ISO standard.