Respiratory hoods, Tychem®, Bullard®
Supplier: BULLARD
20SIC 20SICH 20SICN 20TICH 20TICN 20TICS 20TJ 20TJN RT1
BULL20SICEA 179.53 SGD
BULL20SIC BULL20SICH BULL20SICN BULL20TICH BULL20TICN BULL20TICS BULL20TJ BULL20TJN BULLRT1
Respiratory hoods, Tychem®, Bullard®
Respirators Respiratory Systems
Tychem® hoods provide a variety of simple, effective, and economical options for respiratory protection.
  • Simple, effective, and economical options for respiratory protection
  • Covers entire head, face, and neck to provide splash and overspray protection
  • Neck cuff design maintains positive pressure inside the hood by inflating the inner collar to provide a snug, comfortable fit and continuous airflow
  • Wide-angle lens offers large field of vision while accommodating beards, prescription eyeglasses, or safety glasses
  • Lighter and more comfortable than tight-fitting full facepiece respirators
  • Durable enough to reuse, yet economical enough to discard after each use
  • Approved for NIOSH for use in Type-C applications
Tychem® hoods provide a variety of simple, effective, and economical options for respiratory protection. The hoods cover the entire head, face, and neck to provide splash and overspray protection. The neck cuff design helps maintain positive pressure inside the hood by inflating the inner collar to provide a snug, comfortable fit and a continuous airflow. Hoods feature a wide angle lens that offers a large field of vision while accommodating beards, prescription eyeglasses, or safety glasses. Hoods are lighter and more comfortable than tight-fitting full facepiece respirators. Depending on the work application, hoods are durable enough to reuse, yet economical enough to discard after each use. They do not require fit testing or extensive recordkeeping. Tychem® QC hoods are poly-coated and are ideal for use with pharmaceutical manufacturing, painting, healthcare applications, and general industry. Tychem® SL hoods all feature taped and sealed seams and are SARANEX® film-coated for use in applications where chemical protection properties are critical. Approved for NIOSH for use in Type C applications.