EVAHL PAPRs (powered air purifying respirators), blower assemblies
Supplier: BULLARD
EVAHL1 EVAHL2
BULLEVAHL1EA 1549.75 SGD
BULLEVAHL1 BULLEVAHL2
EVAHL PAPRs (powered air purifying respirators), blower assemblies
Respirators Respiratory Systems
The EVAHL is simple, safe, and dependable. The system features one-button operation, a choice of two air speeds, and a single filter design.
  • Certified for hazardous locations (Class I Division 2, Class II Division 2, Class III)
  • Versatile respirator for particulate or chemical environments
  • Lightweight, compact, well-balanced design
  • Run time: 4 to 10 hours
Additionally, the EVAHL offers Active Flow Technology™, which maintains an airflow of 8,5 CFM (high speed), 25/1000 Assigned Protection Factor (depending upon head top), and dual alarms for low flow and low battery. Finally, the EVAHL has a visual battery fuel gauge to check remaining charge at the push of a button, a brushless motor with ten times the life of a brush motor PAPR, and optional genius charger for battery trending analysis.
Certifications: NIOSH Approvals TC-21C-0922 and TC-23C-2765. ANSI/ISA-12.12.01-2007 (CSA Certificate 2510928) Class I and II, Division and Class III, Division 1 and 2 Hazardous (Classifed) Location.
Ordering information: Filters/cartridges must be ordered separately (except for EVAHL4, where a single HE filter is included).