Foam Brick Gel Packs, Gel Pack Refrigerants, Koolit®
Supplier: COLD CHAIN TECHNOLOGIES
301F 313F
CLDC301FEA 209 SGD
CLDC301F CLDC313F
Foam Brick Gel Packs, Gel Pack Refrigerants, Koolit®
Cooling Packs
Formulated from nontoxic materials, these reusable foam brick gel packs maintain the thermal profile of temperature-sensitive products safely and more efficiently than ice.
The puncture-resistant foam brick gel packs retain a uniform shape and are ideal for exact or tight-fitting configurations. Even heat absorption rate allows for greater cooling longevity.