Screw cap culture tubes with PTFE liner, PYREX®
Supplier: Corning
9826-13 9826-16 9826-16X 9826-16XX 9826-20 9826-20X 9826-25 9826-25X
CORN9826-13EA 716 SGD
CORN9826-13 CORN9826-16 CORN9826-16X CORN9826-16XX CORN9826-20 CORN9826-20X CORN9826-25 CORN9826-25X
Screw cap culture tubes with PTFE liner, PYREX®
Tubes Culture Tubes
Similar to Cat. No. 9825, but phenolic cap has a PTFE interface firmly bonded to the glued-in rubber liner, providing a sure and highly inert sealing face.
For phenolic caps, see Cat. No. 9998.