Culture tube, screw cap, PYREX®
Supplier: Corning
9825-13 9825-16 9825-16X 9825-16XX 9825-20 9825-22 9825-25 9825-25X 9825-38 9825-20X
CORN9825-13EA 551 SGD
CORN9825-13 CORN9825-16 CORN9825-16X CORN9825-16XX CORN9825-20 CORN9825-22 CORN9825-25 CORN9825-25X CORN9825-38 CORN9825-20X
Culture tube, screw cap, PYREX®
Tubes Culture Tubes
Furnished with deep-form phenolic caps to facilitate handling and sealing after autoclaving.
Caps are fabricated from a special formula resistant to the effects of temperature and steam in autoclaving. White enamel marking spot allows excellent surface for pencil notations. Pencil marks are easily erased.
For phenolic caps, see Cat. No. 9999.
Ordering information: Choose from tubes with rubber-lined screw caps or fluoropolymer resin-faced rubber-lined screw caps.