Gooch crucibles, low form, with fritted disc, PYREX®
Supplier: Corning
32960-30C 32960-30F 32960-30M
CORN32960-30CEA 44 SGD
CORN32960-30C CORN32960-30F CORN32960-30M
Gooch crucibles, low form, with fritted disc, PYREX®
Crucibles
Particularly adapted to analytical work in cases where precipitates are dried to constant weight at 110 °C.
  • Made of PYREX® borosilicate glass
Also suitable for high temperatures, in which case it is advisable to heat in an electric furnace. In order to avoid strain, crucible should not be removed from furnace until the temperature has dropped to 250 °C. Limit of use, 500 °C.