Safety Cabinets, En Fire - Rated
Supplier: JUSTRITE MANUFACTURING
22601 22603 22605 22607
JUSU22601EA 10920 SGD
JUSU22601 JUSU22603 JUSU22605 JUSU22607
Safety Cabinets, En Fire - Rated
Furniture Storage Cabinets
Safely store and segregate flammables in either 30 or 90-minute Fire-Rated Safety Cabinets.
  • Hybrid-close door action
  • Manual close doors stop in any position, but self-close in the event of a fire to protect contents
  • Cabinets are supplied with 3 adjustable shelves
Certifications: FM, EN 14470-1, EN 14727, NFPA 30, OSHA, and NFPA 1 - 2012 Edition.
Order Now
SPECIFICATIONS
Environmentally Preferable