Sure-Grip® EX Acid/Corrosive Safety Cabinets
Supplier: JUSTRITE MANUFACTURING
896082 899082
JUSU896082EA 6410 SGD
JUSU896082 JUSU899082
Sure-Grip® EX Acid/Corrosive Safety Cabinets
Furniture Storage Cabinets
Specially designed for safely storing acids and corrosives in laboratories.
  • 10 year warranty
Exclusively formulated paint and poly trays resist acids, bases, and solvents.
Certifications: FM-approved. Complies with NFPA Code 30 and OSHA compliance.
Delivery information: Includes 1 adjustable shelf.
Order Now
SPECIFICATIONS
Environmentally Preferable