Hydroquinone monomethyl ether ≥98%

Supplier: Spectrum Chemical
Warning

Synonyms: p-Methoxyphenol, Mequinol, 4-Methoxyphenol, 1-Hydroxy-4-methoxybenzene, 4-Hydroxyanisole

M2280-500GM M2280-2.5KG
SPCMM2280-500GMEA 272 SGD
SPCMM2280-500GM SPCMM2280-2.5KG
Hydroquinone monomethyl ether ≥98%
Hydroquinone monomethyl ether

4-Methoxyphenol is a white, waxy solid with a phenolic, caramel odor. Also known as Mequinol and MeHQ, it is soluble in benzene, acetone, and alcohol. It is used as an antioxidant to inhibit the radical polymerisation of monomers, a phenol in dermatology, and in dyestuffs. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for consumption.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₇H₈O₂
MW: 124,14 g/mol
Boiling Pt: 243 °C (1013 hPa)
Melting Pt: 55…57 °C
Density: 1,55 g/cm³ (20 °C)
Flash Pt: 128 °C
Storage Temperature: Ambient
MDL Number: MFCD00002332
CAS Number: 150-76-5
EINECS: 205-769-8

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR