PTFE Sleeves, 0.05 mm
Supplier: Ace Glass
7643-108 7643-109
ACEG7643-108EA 163 SGD
ACEG7643-108 ACEG7643-109
PTFE Sleeves, 0.05 mm
NS Joint Sleeves and Connectors Joint Sleeves
PTFE sleeves are inserted between standard taper ground glass joints.
Precludes contamination from lubricant or from leaks. They have a wall thickness of 0,05 mm, are inert, usable to 280 °C, and will withstand low to medium vacuum.