o,α-Dimethylbenzyl alcohol 97

Supplier: Thermo Fisher Scientific

Synonyms: 2-(1-Hydroxyethyl)toluene, 2-Methylphenyl methyl carbinol, α,2-Dimethylbenzyl alcohol, 1-(2-Methylphenyl)ethanol, 1-(o-Tolyl)ethanol

L08065.06 L08065.14
ALFAL08065.06EA 0 SGD
ALFAL08065.06 ALFAL08065.14
o,α-Dimethylbenzyl alcohol 97
o,α-Dimethylbenzyl alcohol

Purity: 97% CAS No.: 7287-82-3 Molecular Formula: C9H12O Formula Weight: 136.20 Refractive Index: 1.5315 Boiling Point: 78-80°/2mm MDL No.: MFCD00046632

Formula: C₉H₁₂O
MW: 136.19
Boiling Pt: 78…80 °C (2 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00046632
CAS Number: 7287-82-3
EINECS: 230-716-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR