(R)-(-)-Methyl mandelate ≥99%

Supplier: Thermo Scientific

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

L14065.03 L14065.06
ALFAL14065.03EA 0 SGD
ALFAL14065.03 ALFAL14065.06
(R)-(-)-Methyl mandelate ≥99%
(R)-(-)-Methyl mandelate

Purity: 99% CAS No.: 20698-91-3 Molecular Formula: C9H10O3 Formula Weight: 166.18 Flash Point: 110°(230°F) Melting Point: 56-58° Beilstein Reference: 3589447 Optical Rotation: -144° (c=1 in methanol) MDL No.: MFCD00064247

Formula: C₉H₁₀O₃
MW: 166,18 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 56…58 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064247
CAS Number: 20698-91-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR