4-Amino-3,5-diiodobenzoic acid tech. 90%, Technical Grade
Supplier: Thermo Scientific
|
Warning
|
A10617.06
A10617.14
ALFAA10617.06EA
0
SGD
ALFAA10617.06
ALFAA10617.14
4-Amino-3,5-diiodobenzoic acid tech. 90%, Technical Grade
4-Amino-3,5-diiodobenzoic acid
Purity: 90%
CAS No.: 2122-61-4
Molecular Formula: C7H5I2NO2
Formula Weight: 388.93
Melting Point:
Formula:
C₇H₅I₂NO₂ MW: 388,93 g/mol Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007684 CAS Number: 2122-61-4 EINECS: 218-331-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...