4-Chloro-4'-fluorobutyrophenone 96%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Chloro-1-(4-fluorophenyl)butan-1-one, ω-Chloro-4-fluorobutyrophenone, γ-Chloro-4-fluorobutyrophenone

A11121.14 A11121.22
ALFAA11121.14EA 0 SGD
ALFAA11121.14 ALFAA11121.22
4-Chloro-4'-fluorobutyrophenone 96%
4-Chloro-4'-fluorobutyrophenone

Purity: 96% CAS No.: 3874-54-2 Molecular Formula: C10H10ClFO Formula Weight: 200.64 Storage Temperature: 2°C to 8°C Flash Point: 152°(306°F) Refractive Index: 1.5255 Melting Point: 5-6° Boiling Point: 120-122°/0.7mm Beilstein Reference: 608741 MDL No.: MFCD00001007

Formula: C₁₀H₁₀ClFO
MW: 200,64 g/mol
Boiling Pt: 120…122 °C (0,7 mmHg)
Melting Pt: 5…6 °C
Density: 1,23
Flash Pt: 152 °C (306 °F)
MDL Number: MFCD00001007
CAS Number: 3874-54-2
EINECS: 223-395-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR