3-Chloro-N-methylaniline ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
N-Methyl-3-chloroaniline
A12418.03
A12418.06
ALFAA12418.03EA
0
SGD
ALFAA12418.03
ALFAA12418.06
3-Chloro-N-methylaniline ≥97%
3-Chloro-N-methylaniline
Purity: 97%
CAS No.: 7006-52-2
Molecular Formula: C7H8ClN
Formula Weight: 141.60
Flash Point:
Formula:
C₇H₈ClN MW: 141,6 g/mol Boiling Pt: 243…244 °C Density: 1,163 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00052014 CAS Number: 7006-52-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...