(2,3-Dichlorophenoxy)acetic acid ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 2,3-D, 2,3-Dichlorophenoxyacetic acid

A13231.06 A13231.14
ALFAA13231.06EA 0 SGD
ALFAA13231.06 ALFAA13231.14
(2,3-Dichlorophenoxy)acetic acid ≥97%
(2,3-Dichlorophenoxy)acetic acid

Purity: 97% CAS No.: 2976-74-1 Molecular Formula: C8H6Cl2O3 Formula Weight: 221.04 Melting Point: 173-175° Beilstein Reference: 1963475

Formula: (C₆H₃Cl₂)OCH₂COOH
MW: 221,04 g/mol
Boiling Pt: 329 °C (1013 hPa)
Melting Pt: 173…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00004299
CAS Number: 2976-74-1
EINECS: 221-022-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR