Allyltrimethylsilane ≥98%
Supplier: Thermo Scientific
|
Danger
|
A14662.09
A14662.18
A14662.30
ALFAA14662.09EA
0
SGD
ALFAA14662.09
ALFAA14662.18
ALFAA14662.30
Allyltrimethylsilane ≥98%
Allyltrimethylsilane
Purity: 98+%
CAS No.: 762-72-1
Molecular Formula: CH2=CHCH2Si(CH3)3
Formula Weight: 114.27
Flash Point: -10°(14°F)
Refractive Index: 1.4080
Boiling Point: 85-86°
Beilstein Reference: 906755
UN No.: 1993
MDL No.: MFCD00008635
Formula:
H₂CCHCH₂Si(CH₃)₃ MW: 114,26 g/mol Boiling Pt: 85…86 °C Density: 0,717 Flash Pt: –10 °C (14 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008635 CAS Number: 762-72-1 EINECS: 212-104-5 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...