Tris(3-fluorophenyl)phosphine ≥97%
Supplier: Thermo Scientific
|
Warning
|
A15117.03
A15117.06
A15117.14
ALFAA15117.03EA
186
SGD
ALFAA15117.03
ALFAA15117.06
ALFAA15117.14
Tris(3-fluorophenyl)phosphine ≥97%
Tris(3-fluorophenyl)phosphine
Purity: 97%
CAS No.: 23039-94-3
Molecular Formula: C18H12F3P
Formula Weight: 316.27
Melting Point: 60-63°
Beilstein Reference: 2946854
Formula:
C₁₈H₁₂F₃P MW: 316,26 g/mol Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015722 CAS Number: 23039-94-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...