3-(Trifluoromethyl)benzoylacetonitrile ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2-Cyano-3'-(trifluoromethyl)acetophenone, 3-Oxo-3-[3-(trifluoromethyl)phenyl]propanenitrile, 3-(3-(Trifluoromethyl)phenyl)-3-oxopropanenitrile
A15564.03
A15564.06
ALFAA15564.03EA
0
SGD
ALFAA15564.03
ALFAA15564.06
3-(Trifluoromethyl)benzoylacetonitrile ≥97%
3-(Trifluoromethyl)benzoylacetonitrile
Purity: 97%
CAS No.: 27328-86-5
Molecular Formula: C10H6F3NO
Formula Weight: 213.16
Melting Point: 58-63°
Beilstein Reference: 2373428
Formula:
C₁₀H₆F₃NO MW: 213,16 g/mol Melting Pt: 58…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052315 CAS Number: 27328-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...