6-Nitro-1H-indazole ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-Nitroindazole, 6-Nitro-1H-indazole, 6-NINA
A16213.14
A16213.22
A16213.36
ALFAA16213.14EA
0
SGD
ALFAA16213.14
ALFAA16213.22
ALFAA16213.36
6-Nitro-1H-indazole ≥97%
6-Nitro-1H-indazole
Purity: 97%
CAS No.: 7597-18-4
Molecular Formula: C7H5N3O2
Formula Weight: 163.14
Odour: Odourless
Melting Point: 179-182°
Beilstein Reference: 7812
MDL No.: MFCD00005695
Formula:
C₇H₅N₃O₂ MW: 163,14 g/mol Melting Pt: 179…182 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005695 CAS Number: 7597-18-4 EINECS: 231-500-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...