5-Fluoro-2-nitrobenzotrifluoride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Fluoro-2-nitrobenzotrifluoride, α,α,α,5-Tetrafluoro-2-nitrotoluene, 1-Fluoro-4-nitro-3-(trifluoromethyl)benzene
A18780.06
A18780.14
ALFAA18780.06EA
0
SGD
ALFAA18780.06
ALFAA18780.14
5-Fluoro-2-nitrobenzotrifluoride ≥98%
5-Fluoro-2-nitrobenzotrifluoride
Purity: 98+%
CAS No.: 393-09-9
Molecular Formula: C7H3F4NO2
Formula Weight: 209.10
Flash Point: 88°(190°F)
Refractive Index: 1.4600
Melting Point: 22-24°
Boiling Point: 87-88°/2mm
Beilstein Reference: 3304470
UN No.: 2811
MDL No.: MFCD00039741
Formula:
C₇H₃F₄NO₂ MW: 209,1 g/mol Boiling Pt: 87…88 °C (2 mmHg) Melting Pt: 22…24 °C Density: 1,497 Flash Pt: 88 °C (190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039741 CAS Number: 393-09-9 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...