1-Fluoro-4-iodo-3-nitrobenzene ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
4-Fluoro-1-iodo-2-nitrobenzene, 5-Fluoro-2-iodonitrobenzene, 2-Iodo-5-fluoronitrobenzene
A18962.03
A18962.06
ALFAA18962.03EA
0
SGD
ALFAA18962.03
ALFAA18962.06
1-Fluoro-4-iodo-3-nitrobenzene ≥97%
1-Fluoro-4-iodo-3-nitrobenzene
Purity: 97%
CAS No.: 364-77-2
Molecular Formula: C6H3FINO2
Formula Weight: 267.00
Refractive Index: 1.6150
Boiling Point: 110°/4mm
MDL No.: MFCD00153169
Formula:
C₆H₃FINO₂ MW: 267 g/mol Boiling Pt: 110 °C (4 mmHg) Melting Pt: 22,5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153169 CAS Number: 364-77-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...