3,5-Dimethyl-4-iodoisoxazole ≥97%

Supplier: Thermo Scientific

A19911.03 A19911.06
ALFAA19911.03EA 0 SGD
ALFAA19911.03 ALFAA19911.06
3,5-Dimethyl-4-iodoisoxazole ≥97%
3,5-Dimethyl-4-iodoisoxazole

Purity: 97% CAS No.: 10557-85-4 Molecular Formula: C5H6INO Formula Weight: 223.01 Melting Point: 51-55° MDL No.: MFCD00173743

Formula: C₅H₆INO
MW: 223,01 g/mol
Melting Pt: 51…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00173743
CAS Number: 10557-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR