Bis(dimethylamino)dimethylsilane 97%
Supplier: Thermo Scientific
|
Danger
|
B21119.14
B21119.22
ALFAB21119.14EA
0
SGD
ALFAB21119.14
ALFAB21119.22
Bis(dimethylamino)dimethylsilane 97%
Bis(dimethylamino)dimethylsilane
Purity: 97%
CAS No.: 3768-58-9
Molecular Formula: [(CH3)2N]2Si(CH3)2
Formula Weight: 146.31
Flash Point: -7°(19°F)
Refractive Index: 1.4170
Melting Point: -98°
Boiling Point: 128-129°
Beilstein Reference: 1736095
UN No.: 2924
MDL No.: MFCD00008296
Formula:
C₆H₁₈N₂Si MW: 146,31 g/mol Boiling Pt: 128…129 °C Melting Pt: –98 °C Density: 0,807 Flash Pt: –7 °C (19 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008296 CAS Number: 3768-58-9 EINECS: 223-200-1 UN: 2924 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...