3-Bromopropiononitrile 97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
3-Bromopropionitrile
B23998.22
B23998.14
ALFAB23998.22EA
0
SGD
ALFAB23998.22
ALFAB23998.14
3-Bromopropiononitrile 97%
3-Bromopropiononitrile
Purity: 98%
CAS No.: 2417-90-5
Molecular Formula: BrCH2CH2CN
Formula Weight: 133.98
Flash Point: 97°(206°F)
Refractive Index: 1.4810
Boiling Point: 200°
Beilstein Reference: 1738565
UN No.: 3276
MDL No.: MFCD00001951
Formula:
BrH₂CCH₂CN MW: 133,98 g/mol Boiling Pt: 200 °C Density: 1,615 Flash Pt: 97 °C (206 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001951 CAS Number: 2417-90-5 EINECS: 219-332-4 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...