2-Chloro-5-(trifluoromethyl)benzylamine ≥97%
Supplier: Thermo Scientific
|
Danger
|
B24449.03
B24449.06
ALFAB24449.03EA
0
SGD
ALFAB24449.03
ALFAB24449.06
2-Chloro-5-(trifluoromethyl)benzylamine ≥97%
2-Chloro-5-(trifluoromethyl)benzylamine
Purity: 97%
CAS No.: 15996-78-8
Molecular Formula: C8H7ClF3N
Formula Weight: 209.60
Flash Point: 100°(212°F)
Refractive Index: 1.4890
Odour: Amine-like
Boiling Point: 90-94°/13mm
Beilstein Reference: 2835666
UN No.: 2735
MDL No.: MFCD00190123
Formula:
C₈H₇ClF₃N MW: 209,6 g/mol Boiling Pt: 90…94 °C (13 mmHg) Density: 1,330 Flash Pt: 100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00190123 CAS Number: 15996-78-8 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...