1-Isopropylpiperazine ≥98%

Supplier: Thermo Scientific
Danger

B25325.03 B25325.06
ALFAB25325.03EA 0 SGD
ALFAB25325.03 ALFAB25325.06
1-Isopropylpiperazine ≥98%
1-Isopropylpiperazine

Purity: 98% CAS No.: 4318-42-7 Molecular Formula: C7H16N2 Formula Weight: 128.22 Flash Point: 54°(130°F) Refractive Index: 1.4700 Boiling Point: 180-181° UN No.: 2733 MDL No.: MFCD00167971

Formula: C₇H₁₆N₂
MW: 128,22 g/mol
Boiling Pt: 180…181 °C
Density: 0,896
Flash Pt: 54 °C (130 °F)
Storage Temperature: Ambient
MDL Number: MFCD00167971
CAS Number: 4318-42-7
EINECS: 224-345-3
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR