5-Iodo-o-anisidine ≥97%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
5-Iodo-2-methoxyaniline, 2-Amino-4-iodoanisole, 5-Iodo-2-anisidine
B25557.03
B25557.06
ALFAB25557.03EA
0
SGD
ALFAB25557.03
ALFAB25557.06
5-Iodo-o-anisidine ≥97%
5-Iodo-o-anisidine
Purity: 97%
CAS No.: 77770-09-3
Molecular Formula: C7H8INO
Formula Weight: 249.05
Melting Point: 86-87°
MDL No.: MFCD03095407
Formula:
C₇H₈INO MW: 249,05 g/mol Melting Pt: 86…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095407 CAS Number: 77770-09-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...