3,6-Difluorophthalic anhydride ≥97%

Supplier: Thermo Scientific
Warning

Synonyms: 4,7-Difluoroisobenzofuran-1,3-dione

H26215.MC H26215.ME
ALFAH26215.MCEA 90.9 SGD
ALFAH26215.MC ALFAH26215.ME
3,6-Difluorophthalic anhydride ≥97%
3,6-Difluorophthalic anhydride

Purity: 97% CAS No.: 652-40-4 Molecular Formula: C8H2F2O3 Formula Weight: 184.10 Melting Point: 218-221° MDL No.: MFCD00134537

Formula: C₈H₂F₂O₃
MW: 184,1 g/mol
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00134537
CAS Number: 652-40-4
EINECS: None

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR