Methylamine 2 M in methanol

Supplier: Thermo Scientific
Danger

Synonyms: Aminomethane

H26889.AE H26889.AU
ALFAH26889.AEEA 0 SGD
ALFAH26889.AE ALFAH26889.AU
Methylamine 2 M in methanol
Methylamine
Formula: CH₅N
MW: 31,06 g/mol
Boiling Pt: ∼40 °C
Density: 0,785
Flash Pt: 7 °C (45 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008104
CAS Number: 74-89-5
EINECS: 200-820-0
UN: 1992
ADR: 3,II
Merck Index: 14,6014

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR