3-Bromoisonicotinonitrile 95%

Supplier: Thermo Scientific
Warning

H27049.MD H27049.03
ALFAH27049.MDEA 0 SGD
ALFAH27049.MD ALFAH27049.03
3-Bromoisonicotinonitrile 95%
3-Bromoisonicotinonitrile

Purity: 95% CAS No.: 13958-98-0 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Odour: Odourless Melting Point: 92-94° MDL No.: MFCD00234143

Formula: C₆H₃BrN₂
MW: 183,01 g/mol
Melting Pt: 92…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00234143
CAS Number: 13958-98-0
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR